missing translation for 'onlineSavingsMsg'
Learn More

rac Didemethyl Citalopram-d6 Hydrochloride, TRC

Product Code. 30370956
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370956 1 mg
30383307 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30370956 Supplier Toronto Research Chemicals Supplier No. TRCD4394771MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189865-88-0
Molecular Formula C18H12D6ClFN2O
Molecular Weight (g/mol) 338.84
SMILES Cl.[2H]C([2H])(N)C([2H])([2H])C([2H])([2H])C1(OCc2cc(ccc12)C#N)c3ccc(F)cc3

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H17FN2O.ClH/c19-16-5-3-15(4-6-16)18(8-1-9-20)17-7-2-13(11-21)10-14(17)12-22-18;/h2-7,10H,1,8-9,12,20H2;1H/i1D2,8D2,9D2;
Chemical Name or Material rac Didemethyl Citalopram-d6 Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.