missing translation for 'onlineSavingsMsg'
Learn More

rac Didemethyl Citalopram Hydrochloride, TRC

Product Code. 30008333
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008333 25 mg 25mg
30005116 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008333 Supplier Toronto Research Chemicals Supplier No. TRCD43947625MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189694-81-2
Molecular Formula C18 H17 F N2 O . H Cl
Molecular Weight (g/mol) 332.8
Synonym 5-Isobenzofurancarbonitrile, 1-(3-aminopropyl)-1-(4-fluorophenyl)-1,3-dihydro-, hydrochloride (1:1), 1-(3-Aminopropyl)-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile hydrochloride, 1-(3-Aminopropyl)-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile hydrochloride, Didemethylcitalopram hydrochloride, Didesmethylcitalopram hydrochloride, Dinorcitalopram hydrochloride, Lu 11-161 HCl, N-Desmethylcitalopram hydrochloride, rac-Didesmethylcitalopram hydrochloride, N,N-Didesmethyl citalopram hydrochloride, N,N-Didesmethylcitalopram hydrochloride, rac Didemethyl citalopram hydrochloride
IUPAC Name 1-(3-aminopropyl)-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrochloride
SMILES Cl.NCCCC1(OCc2cc(ccc12)C#N)c3ccc(F)cc3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H17FN2O.ClH/c19-16-5-3-15(4-6-16)18(8-1-9-20)17-7-2-13(11-21)10-14(17)12-22-18;/h2-7,10H,1,8-9,12,20H2;1H
Formula Weight 332.109
Chemical Name or Material rac Didemethyl Citalopram Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.