missing translation for 'onlineSavingsMsg'
Learn More

rac Ibuprofen (1mg/ml in Methanol), TRC

Product Code. 30389879
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389879 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389879 Supplier Toronto Research Chemicals Supplier No. TRCKIT25855X1ML
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 15687-27-1
Molecular Formula C13 H18 O2
Quantity 5 x 1 mL
Synonym alpha-Methyl-4-(2-methylpropyl)-benzeneacetic acid, p-Isobutylhydratropic acid, (+/-)-alpha-Methyl-4-(2-methylpropyl)benzeneacetic acid, (+/-)-2-(p-Isobutylphenyl)propionic acid, (+/-)-Ibuprofen, (+/-)-Ibuprophen, (4-Isobutylphenyl)-alpha-methyl-acetic acid, (RS)-Ibuprofen, alpha-(4-Isobutylphenyl)propionic acid, alpha-Methyl-4-(2-methylpropyl)benzeneacetic acid, 2-(4'-Isobutylphenyl)propionic acid, 2-(4-Isobutylphenyl)propanoic acid, 2-(p-Isobutylphenyl)propionic acid, 4-Isobutyl-α-methylphenylacetic acid, 4-Isobutylhydratropic acid, Act 3, Actiprofen, Adex 200, Adran, Advil, Alaxan, Algi-Flanderil, Algiflex, Algofen, Am-Fam 400, Amibufen, Anafen, Anco, Andran, Anflagen, Antarene, Antiflam, Apo-Ibuprofen, Apsifen, Artofen, Artril, Artril 300, Atril 300, Balkaprofen, Betaprofen, Bloom, Bluton, Brofen, Brufanic, Brufen, Brufen 400, Brufen Retard, Bruflam, Brufort, Buburone, Buluofen, Burana, Butacortelone, Butylenin, Carol, Cobo, Codral Period Pain, Combiflam, Dansida, Dentigoa, Dibufen, Dolgin, Dolgirid, Dolgit, Dolmaral, Dolo-Dolgit, Dolocyl, Dolofen, Dolofen F, Dolomax, Donjust B, Doretrim, Dorival, Drin, Easifon, Ebufac, Emflam, Emflam 200, Emodin, Epobron, Femadon, Fenbid, Fenspan, Focus, Gofen, Gynofug, Haltran, IB 100, IP 82, Ibosure, Ibren, Ibu-Attritin, Ibu-Tab, Ibu-slow, Ibufen, Ibuflamar, Ibufug, Ibugen, Ibugesic, Ibuleve, Ibulgan, Ibumetin, Ibupirac, Ibupril, Ibuprocin, Ibuprofen, Ibuprohm, Ibusal, Ibutad, Ifen, Inabrin, Inflam, Inza, Ipren, Iprogel, Irfen, Isodol, Lamidon, Librofem, Lidifen, Liptan, Lopane, Mensoton, Motrin, Motrin IB, Mynosedin, NSC 256857, Nagifen-D, Napacetin, Nobafon, Nobfelon, Nobgen, Noritis, Norton, Novogent, Novoprofen, Nuprin, Nurofen, Optifen, Opturem, Ostarin, Ostofen, Paduden, Panafen, Pantrop, Paxofen, Pediaprofen, Perofen, Proartinal, Profen, Proflex, Prontalgin, Provon, Quadrax, RD 13621, Rafen, Ranofen, Recidol, Relcofen, Roidenin, Rufin, Rupan, Seclodin, Stelar, Suspren, Syntofene, Tabalon, Tabalon 400, Tarein, Tatanal, Tofen, Trendar, U 18753, Unipron, Upfen, Uprofen, Urem, Zafen, Zofen, dl-Ibuprofen, p-Isobutyl-2-phenylpropionic acid, p-Isobutylhydratropic acid
InChI Formula InChI=1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)
SMILES CC(C)Cc1ccc(cc1)C(C)C(=O)O
IUPAC Name 2-[4-(2-methylpropyl)phenyl]propanoic acid
Molecular Weight (g/mol) 206.28
Formula Weight 206.1307
Concentration or Composition (by Analyte or Components) Ibuprofen 1 mg/mL
Chemical Name or Material Ibuprofen
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.