missing translation for 'onlineSavingsMsg'
Learn More

rac-Ibuprofen Amide, TRC

Product Code. 30138079
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138079 1 g 1g
30134141 250 mg 250mg
17923005 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138079 Supplier Toronto Research Chemicals Supplier No. TRCI1400351G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59512-17-3
Molecular Formula C13 H19 N O
Molecular Weight (g/mol) 205.3
Synonym (2RS)-2-[4-(2-Methylpropyl)phenyl]propanamide, Ibuprofen Imp. C (EP)
IUPAC Name 2-[4-(2-methylpropyl)phenyl]propanamide
SMILES CC(C)Cc1ccc(cc1)C(C)C(=O)N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H19NO/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H2,14,15)
Formula Weight 205.1467
Percent Purity >95%
Chemical Name or Material (2RS)-2-[4-(2-Methylpropyl)phenyl]propanamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.