CAS RN 59512-17-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
rac-Ibuprofen Amide, TRC
CAS: 59512-17-3 Molecular Formula: C13 H19 N O Molecular Weight (g/mol): 205.3 Synonym: (2RS)-2-[4-(2-Methylpropyl)phenyl]propanamide,Ibuprofen Imp. C (EP) IUPAC Name: 2-[4-(2-methylpropyl)phenyl]propanamide SMILES: CC(C)Cc1ccc(cc1)C(C)C(=O)N
2
(2RS)-2-[4-(2-Methylpropyl)phenyl]propanamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.