missing translation for 'onlineSavingsMsg'
Learn More

rac Ibuprofen, TRC

Product Code. 18083540
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
10g
2.50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18083540 2.5 g 2.50g
17989639 10 g 10g
18004040 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18083540 Supplier Toronto Research Chemicals Supplier No. TRCI1400002.5G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15687-27-1
Molecular Formula C13 H18 O2
Molecular Weight (g/mol) 206.28
Synonym alpha-Methyl-4-(2-methylpropyl)-benzeneacetic acid, p-Isobutylhydratropic acid, (+/-)-alpha-Methyl-4-(2-methylpropyl)benzeneacetic acid, (+/-)-2-(p-Isobutylphenyl)propionic acid, (+/-)-Ibuprofen, (+/-)-Ibuprophen, (4-Isobutylphenyl)-alpha-methyl-acetic acid, (RS)-Ibuprofen, alpha-(4-Isobutylphenyl)propionic acid, alpha-Methyl-4-(2-methylpropyl)benzeneacetic acid, 2-(4'-Isobutylphenyl)propionic acid, 2-(4-Isobutylphenyl)propanoic acid, 2-(p-Isobutylphenyl)propionic acid, 4-Isobutyl-α-methylphenylacetic acid, 4-Isobutylhydratropic acid, Act 3, Actiprofen, Adex 200, Adran, Advil, Alaxan, Algi-Flanderil, Algiflex, Algofen, Am-Fam 400, Amibufen, Anafen, Anco, Andran, Anflagen, Antarene, Antiflam, Apo-Ibuprofen, Apsifen, Artofen, Artril, Artril 300, Atril 300, Balkaprofen, Betaprofen, Bloom, Bluton, Brofen, Brufanic, Brufen, Brufen 400, Brufen Retard, Bruflam, Brufort, Buburone, Buluofen, Burana, Butacortelone, Butylenin, Carol, Cobo, Codral Period Pain, Combiflam, Dansida, Dentigoa, Dibufen, Dolgin, Dolgirid, Dolgit, Dolmaral, Dolo-Dolgit, Dolocyl, Dolofen, Dolofen F, Dolomax, Donjust B, Doretrim, Dorival, Drin, Easifon, Ebufac, Emflam, Emflam 200, Emodin, Epobron, Femadon, Fenbid, Fenspan, Focus, Gofen, Gynofug, Haltran, IB 100, IP 82, Ibosure, Ibren, Ibu-Attritin, Ibu-Tab, Ibu-slow, Ibufen, Ibuflamar, Ibufug, Ibugen, Ibugesic, Ibuleve, Ibulgan, Ibumetin, Ibupirac, Ibupril, Ibuprocin, Ibuprofen, Ibuprohm, Ibusal, Ibutad, Ifen, Inabrin, Inflam, Inza, Ipren, Iprogel, Irfen, Isodol, Lamidon, Librofem, Lidifen, Liptan, Lopane, Mensoton, Motrin, Motrin IB, Mynosedin, NSC 256857, Nagifen-D, Napacetin, Nobafon, Nobfelon, Nobgen, Noritis, Norton, Novogent, Novoprofen, Nuprin, Nurofen, Optifen, Opturem, Ostarin, Ostofen, Paduden, Panafen, Pantrop, Paxofen, Pediaprofen, Perofen, Proartinal, Profen, Proflex, Prontalgin, Provon, Quadrax, RD 13621, Rafen, Ranofen, Recidol, Relcofen, Roidenin, Rufin, Rupan, Seclodin, Stelar, Suspren, Syntofene, Tabalon, Tabalon 400, Tarein, Tatanal, Tofen, Trendar, U 18753, Unipron, Upfen, Uprofen, Urem, Zafen, Zofen, dl-Ibuprofen, p-Isobutyl-2-phenylpropionic acid, p-Isobutylhydratropic acid
IUPAC Name 2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)Cc1ccc(cc1)C(C)C(=O)O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)
Formula Weight 206.1307
Percent Purity >95%
Chemical Name or Material Ibuprofen
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.