missing translation for 'onlineSavingsMsg'
Learn More

rac Indapamide, TRC

Product Code. 30138128
Change view
Click to view available options
Quantity:
50 mg
5 g
10 g
Unit Size:
10g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138128 10 g 10g
17902385 50 mg 50mg
17954235 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138128 Supplier Toronto Research Chemicals Supplier No. TRCI50010010G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26807-65-8
Molecular Formula C16 H16 Cl N3 O3 S
Molecular Weight (g/mol) 365.83
Synonym Indapamide, Benzamide, 3-(aminosulfonyl)-4-chloro-N-(2,3-dihydro-2-methyl-1H-indol-1-yl)- (9CI), Benzamide, 4-chloro-N-(2-methyl-1-indolinyl)-3-sulfamoyl- (8CI), (±)-Indapamide, Bajaten, Damide, Flubest, Fludex, Fludin, Flupamid, Indaflex, Indamide, Indamol, Ipamix, Lozol, Natrilix, Noranat, Racemic indapamide, S 1520, SE 1520, Tandix, Tertensif, Veroxil
IUPAC Name 4-chloro-N-(2-methyl-2,3-dihydroindol-1-yl)-3-sulfamoylbenzamide
SMILES CC1Cc2ccccc2N1NC(=O)c3ccc(Cl)c(c3)S(=O)(=O)N

Specifications

Quantity 10 g
InChI Formula InChI=1S/C16H16ClN3O3S/c1-10-8-11-4-2-3-5-14(11)20(10)19-16(21)12-6-7-13(17)15(9-12)24(18,22)23/h2-7,9-10H,8H2,1H3,(H,19,21)(H2,18,22,23)
Formula Weight 365.0601
Chemical Name or Material Indapamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.