missing translation for 'onlineSavingsMsg'
Learn More

rac N,O-Didesmethyl Tramadol, TRC

Product Code. 30012501
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012501 1 mg 1mg
30006565 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012501 Supplier Toronto Research Chemicals Supplier No. TRCD4412201MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138853-73-3
Molecular Formula C14 H21 N O2
Molecular Weight (g/mol) 235.32
Synonym Phenol, 3-[(1R,2R)-1-hydroxy-2-[(methylamino)methyl]cyclohexyl]-, rel-, Phenol, 3-[1-hydroxy-2-[(methylamino)methyl]cyclohexyl]-, cis-(±)-, rel-3-[(1R,2R)-1-Hydroxy-2-[(methylamino)methyl]cyclohexyl]phenol, Phenol, 3-[1-hydroxy-2-[(methylamino)methyl]cyclohexyl]-, cis-, N,O-Didesmethyltramadol, rac N,O-Didesmethyl Tramadol, 3-[(1RS,2RS)-1-Hydroxy-2-[(methylamino)methyl]cyclohexyl]phenol, cis-3-[1-Hydroxy-2-[(methylamino)methyl]cyclohexyl]phenol
IUPAC Name 3-[(1R,2R)-1-hydroxy-2-(methylaminomethyl)cyclohexyl]phenol
SMILES CNC[C@H]1CCCC[C@]1(O)c2cccc(O)c2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C14H21NO2/c1-15-10-12-5-2-3-8-14(12,17)11-6-4-7-13(16)9-11/h4,6-7,9,12,15-17H,2-3,5,8,10H2,1H3/t12-,14+/m1/s1
Formula Weight 235.1572
Chemical Name or Material (+/-)-N,O-Di-Desmethyl Tramadol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.