missing translation for 'onlineSavingsMsg'
Learn More

rac Naringenin, TRC

Product Code. 30133675
Change view
Click to view available options
Quantity:
1 g
5 g
500 g
Unit Size:
1g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133675 500 g 500g
17964335 1 g 1g
17950515 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133675 Supplier Toronto Research Chemicals Supplier No. TRCN378950500G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67604-48-2
Molecular Formula C15 H12 O5
Molecular Weight (g/mol) 272.25
Synonym rac-Naringenin, 2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, (R,S)-Naringenin, (RS)-Naringenin, (±)-5,7,4'-Trihydroxyflavanone, (±)-Naringenin, 4',5,7-Trihydroxyflavanone, 5,7,4'-Trihydroxyflavanone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-chroman-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chroman-4-one, ST 057236, 4H-1-Benzopyran-4-one, 2,3-dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-
IUPAC Name 5,7-dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
SMILES Oc1ccc(cc1)C2CC(=O)c3c(O)cc(O)cc3O2

Specifications

Quantity 500 g
InChI Formula InChI=1S/C15H12O5/c16-9-3-1-8(2-4-9)13-7-12(19)15-11(18)5-10(17)6-14(15)20-13/h1-6,13,16-18H,7H2
Formula Weight 272.0685
Chemical Name or Material omega-Benzoyloxyresacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.