missing translation for 'onlineSavingsMsg'
Learn More

rac Octopamine Hydrochloride, TRC

Product Code. 30133393
Change view
Click to view available options
Quantity:
50 mg
100 mg
5 g
Unit Size:
100mg
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133393 5 g 5g
30006906 50 mg 50mg
30003878 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133393 Supplier Toronto Research Chemicals Supplier No. TRCO2397505G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 770-05-8
Molecular Formula C8 H11 N O2 . Cl H
Molecular Weight (g/mol) 189.64
Synonym Benzenemethanol, α-(aminomethyl)-4-hydroxy-, hydrochloride (9CI), Benzenemethanol, α-(aminomethyl)-4-hydroxy-, hydrochloride, (±)-, Benzyl alcohol, α-(aminomethyl)-p-hydroxy-, hydrochloride, (±)- (8CI), (±)-Octopamine hydrochloride, (±)-p-Octopamaine hydrochloride, 2-Amino-1-(4-hydroxyphenyl)ethanol hydrochloride, 4-(2-Amino-1-hydroxyethyl)phenol hydrochloride, DL-2-Amino-1-(4-hydroxyphenyl)ethanol hydrochloride, DL-Octopamine hydrochloride, Epirenor, NSC 108685, Norfen, Octopamine hydrochloride, dl-Octopamine hydrochloride
IUPAC Name 4-(2-amino-1-hydroxyethyl)phenol;hydrochloride
SMILES Cl.NCC(O)c1ccc(O)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H11NO2.ClH/c9-5-8(11)6-1-3-7(10)4-2-6;/h1-4,8,10-11H,5,9H2;1H
Formula Weight 189.0557
Chemical Name or Material Octopamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.