missing translation for 'onlineSavingsMsg'
Learn More

rac-Synephrine Hydrochloride, TRC

Product Code. 30137699
Change view
Click to view available options
Quantity:
5 mg
10 mg
5 g
Unit Size:
10mg
5g
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137699 5 mg 5mg
30136437 10 mg 10mg
30009605 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137699 Supplier Toronto Research Chemicals Supplier No. TRCS9200015MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5985-28-4
Molecular Formula C9 H13 N O2 . Cl H
Molecular Weight (g/mol) 203.67
Synonym Benzenemethanol, 4-hydroxy-α-[(methylamino)methyl]-, hydrochloride (1:1), Benzenemethanol, 4-hydroxy-α-[(methylamino)methyl]-, hydrochloride (9CI), Benzyl alcohol, p-hydroxy-α-[(methylamino)methyl]-, hydrochloride (8CI), Oxedrine hydrochloride, Synephrine hydrochloride, p-Hydroxy-α-(methylaminomethyl)benzyl alcohol hydrochloride
IUPAC Name 4-[1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
SMILES Cl.CNCC(O)c1ccc(O)cc1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C9H13NO2.ClH/c1-10-6-9(12)7-2-4-8(11)5-3-7;/h2-5,9-12H,6H2,1H3;1H
Formula Weight 203.0713
Chemical Name or Material rac-Synephrine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.