missing translation for 'onlineSavingsMsg'
Learn More

rac,trans-Milnacipran Hydrochloride, TRC

Product Code. 30477122
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477122 50 mg 50mg
30470055 10 mg 10mg
30466536 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477122 Supplier Toronto Research Chemicals Supplier No. TRCM34462550MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105310-47-2
Molecular Formula C15H23ClN2O
Molecular Weight (g/mol) 282.81
Synonym (1R,2R)-rel-2-(Aminomethyl)-N,N-diethyl-1-phenyl-cyclopropanecarboxamide Monohydrochloride, trans-2-(Aminomethyl)-N,N-diethyl-1-phenyl-cyclopropanecarboxamide Monohydrochloride
IUPAC Name (1S,2S)-2-(aminomethyl)-N,N-diethyl-1-phenylcyclopropane-1-carboxamide;hydrochloride
SMILES NC[C@H]1C[C@]1(C2=CC=CC=C2)C(N(CC)CC)=O.Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H22N2O.ClH/c1-3-17(4-2)14(18)15(10-13(15)11-16)12-8-6-5-7-9-12;/h5-9,13H,3-4,10-11,16H2,1-2H3;1H/t13-,15-;/m1./s1
Formula Weight 282.1499
Percent Purity >95%
Chemical Name or Material rac,trans-Milnacipran Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.