Learn More
RAD1 Mouse, anti-Human, Clone: 1A11E2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68637-1-IG-20UL
 View more versions of this product
Product Code. 18048045
  • 145.00 € / 20µL
Description
The RAD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAD1. This antibody recognizes human antigen. The RAD1 antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
Specifications
RAD1
1A11E2
HRAD1, RAD1, RAD1 homolog (S. pombe), REC1
20 μL
5810
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, Immunoprecipitation, ELISA
1000 μg/mL
Mouse
Primary
Human
150 kDa
IgG2a