Learn More
Invitrogen™ RAD23A Recombinant Rabbit Monoclonal Antibody (24GB1080)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557673
 View more versions of this product
Product Code. 30276021
  • 198.00 € / 20µL
Description
RAD23A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The product of this gene is a small iron-sulfur protein that transfers electrons from NADPH through ferredoxin reductase to a terminal cytochrome P450. This particular oxidation/reduction system is found in steroidogenic tissues, and is involved with the synthesis of bile acid and vitamin D. In addition to the expressed gene at this chromosomal locus (11q22), there are pseudogenes located on chromosomes 20 and 21. This gene product has been identified in a number of different tissues but all forms have been shown to be identical and are not tissue specific.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RAD23A
Recombinant Monoclonal
Unconjugated
RAD23A
2310040P19Rik; AL024030; HHR23A; HR23A; MGC111083; Mhr23a; rad23; RAD23 homolog A (S. cerevisiae); RAD23 homolog A, nucleotide excision repair protein; RAD23, yeast homolog, A; RAD23A; RAD23a homolog (S. cerevisiae); UV excision repair protein RAD23 homolog A
Rabbit
Affinity chromatography
RUO
19358, 5886
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1080
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P54725, P54726
RAD23A
A synthesized peptide derived from human RAD23A (aa 205-250).
20 μL
Primary
Human, Mouse