Learn More
Rad51 Antibody (523713), Unconjugated, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals MAB99481
 View more versions of this product
Product Code. 18615544
  • 494.00 € / 100µg
Description
Rad51 Monoclonal antibody specifically detects Rad51 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rad51
Monoclonal
1.0 mg/mL
Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 5 - 10 μg/mL
BRCC5, HRAD51, HsRAD51, HsT16930, RAD51 (S. cerevisiae) homolog (E coli RecA homolog), RAD51 homolog (RecA homolog, E. coli) (S. cerevisiae), RAD51 homolog A, RAD51ABRCA1/BRCA2-containing complex, subunit 5, RAD51L3, RecA, E. coli, homolog of, RECADNA repair protein RAD51 homolog 1, RecA-like protein, recombination protein A
E. coli-derived partial recombinant human Rad51 protein (amino acids 174-339)
100 μg
Cancer, DNA Repair, Homologous Recombination, modENCODE Antibodies
5888
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG3
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
523713
Unconjugated
PBS
Mouse
Protein A or G purified
RUO
Primary
Human
Purified