missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rad51 Antibody (523713), Unconjugated, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB99481-SP
This item is not returnable.
View return policy
Description
Rad51 Monoclonal antibody specifically detects Rad51 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Rad51 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 5 - 10 μg/mL | |
| BRCC5, HRAD51, HsRAD51, HsT16930, RAD51 (S. cerevisiae) homolog (E coli RecA homolog), RAD51 homolog (RecA homolog, E. coli) (S. cerevisiae), RAD51 homolog A, RAD51ABRCA1/BRCA2-containing complex, subunit 5, RAD51L3, RecA, E. coli, homolog of, RECADNA repair protein RAD51 homolog 1, RecA-like protein, recombination protein A | |
| E. coli-derived partial recombinant human Rad51 protein (amino acids 174-339) | |
| 25 μg | |
| Cancer, DNA Repair, Homologous Recombination, modENCODE Antibodies | |
| 5888 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG3 |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 523713 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction