missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD52 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95195-0.1ml
This item is not returnable.
View return policy
Description
RAD52 Polyclonal antibody specifically detects RAD52 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RAD52 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| DNA repair protein RAD52 homolog, RAD52 (S. cerevisiae) homolog, RAD52 homolog (S. cerevisiae), recombination protein RAD52, rhabdomyosarcoma antigen MU-RMS-40.23 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 125-418 of human RAD52 (NP_602296.2). GYGVSEGLKSKALSLEKARKEAVTDGLKRALRSFGNALGNCILDKDYLRSLNKLPRQLPLEVDLTKAKRQDLEPSVEEARYNSCRPNMALGHPQLQQVTSPSRPSHAVIPADQDCSSRSLSSSAVESEATHQRKLRQKQLQQQFRERMEKQQVRVSTPSAEKSEAAPPAPPVTHSTPVTVSEPLLEKDFLAGVTQELIKTLEDNSEKWAVTPDAGDGVVKPSSRADPAQTSDTLALNNQMVTQNRTPHSVCHQKPQAKSGSWDLQTYSADQRTTGNWESHRKSQDMKKRKYDPS | |
| 0.1 mL | |
| Breast Cancer, DNA Repair, Homologous Recombination | |
| 5893 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction