missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD52 Antibody (SR1844), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22254-100ul
This item is not returnable.
View return policy
Description
RAD52 Monoclonal antibody specifically detects RAD52 in Human samples. It is validated for Western Blot
Specifications
| RAD52 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1844 | |
| Western Blot 1:500-1:2000 | |
| DNA repair protein RAD52 homolog, RAD52 (S. cerevisiae) homolog, RAD52 homolog (S. cerevisiae), recombination protein RAD52, rhabdomyosarcoma antigen MU-RMS-40.23 | |
| A synthesized peptide derived from human RAD52 (Uniprot #: P43351) | |
| 100 μg | |
| Breast Cancer, DNA Repair, Homologous Recombination | |
| 5893 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction