missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD54B Polyclonal Antibody

Description
RAD54B Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | RAD54B |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | RAD54B |
| Gene Accession No. | Q9Y620 |
| Gene Alias | DNA repair and recombination protein RAD54B; E130016E03Rik; fibrinogen silencer-binding protein; FSBP; LOW QUALITY PROTEIN: DNA repair and recombination protein RAD54B; RAD54 homolog B; RAD54 homolog B (S. cerevisiae); RAD54, S. cerevisiae, homolog of, B; Rad54b; RDH54; RGD1306507 |
| Gene Symbols | RAD54B |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?