missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD9B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06241-100ul
This item is not returnable.
View return policy
Description
RAD9B Polyclonal antibody specifically detects RAD9B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RAD9B | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:5000 | |
| cell cycle checkpoint control protein RAD9B, cell cycle checkpoint control protein RAD9B homolog, DNA repair exonuclease rad9 homolog B, FLJ40346, hRAD9B, MGC75426, RAD9 homolog B (S. pombe) | |
| Produced in rabbits immunized with E. coli-derived Human RAD9B fragment. | |
| 100 μg | |
| Cell Cycle and Replication | |
| 144715 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur