missing translation for 'onlineSavingsMsg'
Learn More

Radezolid, MedChemExpress

Product Code. 16438777
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438777 5 mg 5mg
16436178 10 mg 10mg
16402317 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16438777 Supplier Medchem Express Supplier No. HY14800
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Radezolid (RX-1741) is a oxazolidinone antibiotic. Radezolid is active against Staphylococcus, Chlamydia, and Legionella species, and remains active against Linezolid-resistant strains.

Radezolid (RX-1741) is a oxazolidinone antibiotic. Radezolid is active against Staphylococcus, Chlamydia, and Legionella species, and remains active against Linezolid-resistant strains.

Specifications

Chemical Name or Material Radezolid
Physical Form Solid
CAS 869884-78-6
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H23FN6O3
For Use With (Application) COVID-19-immunoregulation
Synonym RX-1741
Solubility Information DMSO : ≥ 25 mg/mL (57.02 mM)
SMILES CC(NC[C@H]1CN(C2=CC=C(C3=CC=C(CNCC4=CN=NN4)C=C3)C(F)=C2)C(O1)=O)=O
Molecular Weight (g/mol) 438.45
Formula Weight 438.45
Percent Purity 98.02%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.