Learn More
Invitrogen™ RAE1 alpha/beta/delta/epsilon Monoclonal Antibody (199205)
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  Invitrogen™ MA561040
Product Code. 30533643
  • 589.00 € / 500µg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
RAE1 (RNA Export 1) is a gene that plays a pivotal role in the nuclear export of mRNA in eukaryotic cells. This gene encodes a protein that is integral to the mRNA export machinery, ensuring the proper transport of mRNA from the nucleus to the cytoplasm for translation. The gene has several isoforms, including RAE1 alpha, beta, delta, and epsilon, which may have unique or overlapping functions in different tissues or developmental stages. The proper functioning of RAE1 and its isoforms is crucial for maintaining normal cellular processes and gene expression. Disruptions or mutations in the RAE1 gene can lead to significant defects in mRNA export, potentially causing a range of diseases, including neurodevelopmental disorders and cancers. Ongoing research into RAE1 and its various isoforms is essential for a deeper understanding of mRNA transport mechanisms and their broader implications in cellular health and disease.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RAE1 alpha/beta/delta/epsilon
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
BaF3 mouse pro-B cell line transfected with Rae-1 beta (Accession # NP_033043) and Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta Leu29-Ser227 (Accession # Q9JI58).
500 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Flow Cytometry, Inhibition Assays
199205
Unconjugated
Rat
Protein A/G
RUO
Mouse
Antibody
IgG2a