missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAG1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 470.00 €
Specifications
| Antigen | RAG1 |
|---|---|
| Dilution | Western Blot 1:1000-1:3000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18695220
|
Novus Biologicals
NBP2-93122-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665310
|
Novus Biologicals
NBP2-93122-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAG1 Polyclonal antibody specifically detects RAG1 in Mouse samples. It is validated for Western BlotSpecifications
| RAG1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Direct Reversal of DNA Damage, Immunology | |
| PBS (pH 7.3), 50% glycerol | |
| 5896 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:3000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| MGC43321, RAG-1, recombination activating gene 1, RING finger protein 74, RNF74recombination activating protein 1, V(D)J recombination-activating protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human RAG1 (NP_000439.1). MAASFPPTLGLSSAPDEIQHPHIKFSEWKFKLFRVRSFEKTPEEAQKEKKDSFEGKPSLEQSPAVLDKADGQKPVPTQPLLKAHPKFSKKFHDNEKARGKAIHQANLRHLCRICGNSFRADEHNRRYPVHGPVDGKTLGLLRKKEKRATSWPDLIAKVFRIDVKADVDSIHPTEFCHNCWSIMHRKFSSAPCEVYFPRNVTMEWHPHTPSCDICNTARRGLKRKSLQPNLQLSKKLKTVLDQARQARQRKRRAQARISSKDVMKKIANCS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title