missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ragifilimab Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

477.00 € - 1204.00 €

Spécification

Antigen Ragifilimab Humanized
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantity Prix Quantité et disponibilité  
Code produit Marque Quantity Prix Quantité et disponibilité  
30284125
GREENER_CHOICE
Afficher les documents
Invitrogen™
MA558597
100 μg
477.00 €
100µg
Voir les produits de remplacement
Économisez jusqu'à 
»
Please to purchase this item. Need a web account? Register with us today!
30284119
Afficher les documents
Invitrogen™
MA558598
1 mg
1204.00 €
1mg
Voir les produits de remplacement
Économisez jusqu'à 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Ragifilimab is an anti-LAG-3 monoclonal antibody that inhibits the LAG-3 pathway, enhancing T-cell activation and immune response against tumors.
Specifications

Specifications

Ragifilimab Humanized
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.