missing translation for 'onlineSavingsMsg'
Learn More

Ralimetinib dimesylate, MedChemExpress

Product Code. 16495578
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495578 10 mg 10mg
16478277 5 mg 5mg
16466126 100 mg 100mg
16461747 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495578 Supplier Medchem Express Supplier No. HY13241
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ralimetinib dimesylate (LY2228820 dimesylate) is a selective, ATP-competitive inhibitor of p38 MAPK α/β with IC50s of 5.3 and 3.2 nM, respectively. Ralimetinib (LY2228820) selectively inhibits phosphorylation of MK2 (Thr334), with no effect on phosphorylation of p38a MAPK, JNK, ERK1/2, c-Jun, ATF2, or c-Myc.

Ralimetinib dimesylate (LY2228820 dimesylate) is a selective, ATP-competitive inhibitor of p38 MAPK α/β with IC50s of 5.3 and 3.2 nM, respectively. Ralimetinib (LY2228820) selectively inhibits phosphorylation of MK2 (Thr334), with no effect on phosphorylation of p38a MAPK, JNK, ERK1/2, c-Jun, ATF2, or c-Myc.

Specifications

Chemical Name or Material Ralimetinib dimesylate
CAS 862507-23-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H37FN6O6S2
Quantity 10 mg
Synonym LY2228820 dimesylate
Solubility Information DMSO : 61 mg/mL (99.55 mM; Need ultrasonic and warming)
SMILES FC1=CC=C(C=C1)C2=C(NC(C(C)(C)C)=N2)C3=NC4=C(C=C3)N=C(N)N4CC(C)(C)C.OS(C)(=O)=O.OS(C)(=O)=O
Molecular Weight (g/mol) 612.74
Formula Weight 612.74
Percent Purity 96.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.