missing translation for 'onlineSavingsMsg'
Learn More

Ralinepag, MedChemExpress

Product Code. 16457178
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16457178

Brand: Medchem Express HY16751

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ralinepag is a potent, orally bioavailable and non-prostanoid prostacyclin (IP) receptor agonist, with EC50s of 8.5 nM, 530 nM and 850 nM for human and rat IP receptor and human DP1 receptor, respectively.

Ralinepag is a potent, orally bioavailable and non-prostanoid prostacyclin (IP) receptor agonist, with EC50s of 8.5 nM, 530 nM and 850 nM for human and rat IP receptor and human DP1 receptor, respectively.

Specifications

Chemical Name or Material Ralinepag
Physical Form Solid
CAS 1187856-49-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C23H26ClNO5
For Use With (Application) COVID-19-immunoregulation
Synonym APD811
Solubility Information DMSO : ≥ 100 mg/mL (231.53 mM)
SMILES O=C(O)COC[C@H]1CC[C@H](COC(N(C2=CC=C(Cl)C=C2)C3=CC=CC=C3)=O)CC1
Molecular Weight (g/mol) 431.91
Formula Weight 431.91
Percent Purity 99.7%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.