missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ralinepag, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Ralinepag is a potent, orally bioavailable and non-prostanoid prostacyclin (IP) receptor agonist, with EC50s of 8.5 nM, 530 nM and 850 nM for human and rat IP receptor and human DP1 receptor, respectively.
Specifications
Specifications
| Chemical Name or Material | Ralinepag |
| Physical Form | Solid |
| CAS | 1187856-49-0 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C23H26ClNO5 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | APD811 |
| Solubility Information | DMSO : ≥ 100 mg/mL (231.53 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction