missing translation for 'onlineSavingsMsg'
Learn More

Raltegravir potassium, MedChemExpress

Product Code. 16483858
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483858 10 mg 10mg
16456587 5 mg 5mg
16410237 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16483858 Supplier Medchem Express Supplier No. HY10353A
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Raltegravir (MK 0518) potassium is a potent integrase (IN) inhibitor, used to treat HIV infection.

Raltegravir (MK 0518) potassium is a potent integrase (IN) inhibitor, used to treat HIV infection.

Specifications

Chemical Name or Material Raltegravir potassium
Physical Form Solid
CAS 871038-72-1
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H20FKN6O5
For Use With (Application) COVID-19-anti-virus
Synonym MK 0518 potassium
Solubility Information H2O : 25 mg/mL (51.81 mM; Need ultrasonic) ∣DMSO : 20.83 mg/mL (43.17 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C(N=C(C(NC(C1=NN=C(C)O1)=O)(C)C)N2C)=C(O[K])C2=O)NCC3=CC=C(F)C=C3
Molecular Weight (g/mol) 482.51
Formula Weight 482.51
Percent Purity 97.25%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.