missing translation for 'onlineSavingsMsg'
Learn More

Ramatroban, MedChemExpress

Product Code. 16474467
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474467 50 mg 50mg
16458598 10 mg 10mg
16408446 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16474467 Supplier Medchem Express Supplier No. HYB0745

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ramatroban is a selective thromboxane A2 (TxA2, IC50=14 nM) antagonist, which also antagonizes CRTH2 (IC50=113 nM) by inhibiting PGD2 binding.

Ramatroban is a selective thromboxane A2 (TxA2, IC50=14 nM) antagonist, which also antagonizes CRTH2 (IC50=113 nM) by inhibiting PGD2 binding.

Specifications

Chemical Name or Material Ramatroban
CAS 116649-85-5
Color White
Purity Grade Notes Research
Molecular Formula C21H21FN2O4S
Quantity 50 mg
Synonym BAY u3405
Solubility Information DMSO : 125 mg/mL (300.14 mM; Need ultrasonic)
SMILES O=C(O)CCN1C2=C(C3=C1C=CC=C3)C[C@H](NS(=O)(C4=CC=C(F)C=C4)=O)CC2
Molecular Weight (g/mol) 416.47
Formula Weight 416.47
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.