missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAMP1 Rabbit anti-Human, Mouse, Rat, Clone: S04-2I8, Novus Biologicals™
Rabbit Monoclonal Antibody
286.00 € - 581.00 €
Specifications
| Antigen | RAMP1 |
|---|---|
| Clone | S04-2I8 |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18348392
|
Novus Biologicals
NBP3-15087-25UL |
25 μg |
286.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18305583
|
Bio-Techne
NBP3-15087-100UL |
100 μg |
581.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAMP1 Monoclonal antibody specifically detects RAMP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| RAMP1 | |
| Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| calcitonin receptor-like receptor activity modifying protein 1, Calcitonin-receptor-like receptor activity-modifying protein 1, CRLR activity-modifying protein 1, receptor (calcitonin) activity modifying protein 1, receptor (G protein-coupled) activity modifying protein 1, receptor activity modifying protein 1, receptor activity-modifying protein 1 | |
| A synthetic peptide of human RAMP1 (Uniprot#: O60894) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S04-2I8 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuroscience, Signal Transduction | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 10267 | |
| IgG | |
| Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title