missing translation for 'onlineSavingsMsg'
Learn More

RANBP10 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

190.00 € - 470.00 €

Specifications

Antigen RANBP10
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18620822
View Documents
Novus Biologicals
NBP2-94389-0.02ml
0.02 mL
190.00 €
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18627512
View Documents
Novus Biologicals
NBP2-94389-0.1ml
0.1 mL
470.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RANBP10 Polyclonal antibody specifically detects RANBP10 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

RANBP10
Western Blot
Unconjugated
Rabbit
Human, Mouse
KIAA1464FLJ31165, RAN binding protein 10, ran-binding protein 10, RanBP10
Recombinant fusion protein containing a sequence corresponding to amino acids 362-497 of human RANBP10 (NP_065901.1). YPGSPSLSPRHGPSSSHMHNTGADSPSCSNGVASTKSKQNHSKYPAPSSSSSSSSSSSSSSPSSVNYSESNSTDSTKSQHHSSTSNQETSDSEMEMEAEHYPNGVLGSMSTRIVNGAYKHEDLQTDESSMDDRHPR
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
57610
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.