missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ EvenCoat Goat Anti-Mouse IgG Coated Plates
Click to view available options
Quantity:
15/Pk.
5/Pk.
Unit Size:
Pack of 15
Pack of 5
Specifications
Specifications
| Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
| Surface Treatment | EvenCoat Goat Anti-Mouse IgG |
| For Use With (Application) | ELISA |
| Quantity | 5/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction