Learn More
Invitrogen™ RAP1A/RAP1B Recombinant Rabbit Monoclonal Antibody (24GB1030)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557210
 View more versions of this product
Product Code. 30275551
  • 146.00 € / 20µL
Description
RAP1A/RAP1B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RAP1A/RAP1B
Recombinant Monoclonal
Unconjugated
RAP1A
2810443E11Rik; AI848598; C21KG; G-22K; GTP-binding protein smg p21A; GTP-binding protein smg p21B; GTP-binding protein SMG-P21A; K-REV; KREV1; Krev-1; OK/SW-cl.11; RAL1B; rap 1A; RAP1; RAP1A; RAP-1A; RAP1A, member of RAS oncogene family; Rap1b; RAP1B, member of RAS oncogene family; Ras family small GTP binding protein RAP1B; RAS related protein 1b; RAS-related protein 1a; Ras-related protein Krev-1; Ras-related protein Rap-1A; RAS-related protein RAP1B; ras-related protein Rap-1b; RAS-related protein-1a; small GTP binding protein; SMGP21
Rabbit
Affinity chromatography
RUO
109905, 171337, 215449, 295347, 5906, 5908
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1030
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P61224, P62834, P62835, P62836, Q62636, Q99JI6
RAP1A, RAP1B
A synthesized peptide derived from human RAP1A/RAP1B (aa 100-150).
20 μL
Primary
Human, Mouse, Rat