missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAP80 Recombinant Rabbit Monoclonal Antibody (K01_2P24)
GREENER_CHOICE

Product Code. 30830063
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830063 100 μL
30830387 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830063 Supplier Invitrogen™ Supplier No. MA564188

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

RAP80 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

RAP80 was initially identified as zinc-finger containing nuclear protein that is highly expressed in testis and interacts with the retinoid-related testis-associated receptor (RTR). Later experiments revealed that RAP80 is recruited by the Coiled-coil domain 98 (CCDC98) protein to the breast cancer-1 protein BRCA1, allowing the formation of BRCA1 foci in response to DNA damage caused by ionizing radiation. Both RAP80 and CCDC98 are required for DNA damage resistance, G2-M checkpoint control, and DNA repair. Cells depleted of either RAP80 or CCDC98 exhibited increased sensitivity to ionizing radiation, although not as much as in BRCA1-depleted cells, suggesting that RAP80 and CCDC98 control only part of the DNA damage response role of BRCA1. At least four isoforms of RAP80 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP80
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2P24
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene UIMC1
Gene Accession No. Q96RL1
Gene Alias 9430016E08Rik; BRCA1-A complex subunit RAP80; D330018D10Rik; D630032M02Rik; RAP80; Receptor-associated protein 80; retinoid X receptor interacting protein 110; retinoid X receptor-interacting protein 110; RGD1307009; RIP110; Rxrip110; ubiquitin interaction motif containing 1; ubiquitin interaction motif-containing protein 1; Uimc1; X2HRIP110
Gene Symbols UIMC1
Host Species Rabbit
Immunogen A synthetic peptide of human RAP80.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51720
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.