missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Rapamycin

Product Code. 15682517
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15682517 1 mg 1mg
17130212 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15682517 Supplier STEMCELL Technologies Supplier No. 73362
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antibiotic; mTOR pathway inhibitor; Inhibits FKBP-12

  • Rapamycin is a macrolide antibiotic and immunosuppressive compound that inhibits mammalian target of rapamycin (mTOR) signaling
  • It acts through formation of a complex with cytosolic FK-binding protein 12 (FKBP-12), which directly binds to mTOR complex 1 (mTORC1)
  • Its immunosuppressive effects are mediated through inhibition of IL-2 signaling that is critical for T-cell proliferation and activation (Gibbons et al.; Kay et al.) Rapamycin shows antifungal activity against Candida albicans and other fungi (Vézina et al.)
  • Inhibits growth of MDA-MB-468 human breast cancer cells in vitro, and inhibits tumor growth in a mouse xenograft model in vivo (Akcakanat et al.)
  • Induces autophagy in malignant glioma cells (Takeuchi et al.)

Specifications

CAS 53123-88-9
Quantity 1 mg
Percent Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.