missing translation for 'onlineSavingsMsg'
Learn More

Melford Rapamycin

Product Code. 30018764
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018764 5 mg 5mg
30018763 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018764 Supplier Melford Supplier No. R645000.005
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapamycin, a powerful compound derived from Streptomyces hygroscopicus, has become a cornerstone in the life sciences field due to its versatile applications and pivotal role in understanding cellular and molecular processes.

Rapamycin, a powerful compound derived from Streptomyces hygroscopicus, has become a cornerstone in the life sciences field due to its versatile applications and pivotal role in understanding cellular and molecular processes. In cell biology and molecular research, Rapamycin is renowned for its capacity to selectively inhibit the mTOR (mammalian target of rapamycin) pathway. This pathway regulates fundamental cellular functions, including growth, metabolism, and autophagy, making Rapamycin an indispensable tool for dissecting intricate cellular mechanisms.

In cancer research, Rapamycin has revolutionized the field by targeting mTOR to halt the uncontrolled proliferation of cancer cells, offering promising avenues for novel cancer therapies. Rapamycin's multifaceted nature and various forms ensure its continued prominence in the life sciences, facilitating breakthroughs in cell biology, transplantation medicine, and cancer research.

Specifications

Chemical Name or Material Rapamycin
CAS 53123-88-9
Quantity 5 mg
Molecular Formula C51H79NO13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.