missing translation for 'onlineSavingsMsg'
Learn More

Rapamycin, 99.0%, MP Biomedicals™

Product Code. 11471431 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471431 1 mg 1mg
11461431 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471431 Supplier MP Biomedicals Supplier No. 0215934691
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 53123-88-9
Molecular Formula C51H79NO13
Molecular Weight (g/mol) 914.187
InChI Key QFJCIRLUMZQUOT-HPLJOQBZSA-N
Synonym A422989, NSC226080
PubChem CID 5284616
ChEBI CHEBI:9168
SMILES CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)O)C)C)O)OC)C)C)C)OC

Specifications

Type Rapamycin
Physical Form Powder
Color White
Quantity 1 mg
Assay Percent Range 0.99
Formula Weight 914.2
Percent Purity ≥98%
Grade Not Available
Chemical Name or Material Rapamycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.