Learn More
Neogen Petrifilm™ Rapid Coliform Count Plate
Product Code. 17203754
Encompass
Change view
Click to view available options
Quantity:
50
500
Unit Size:
Pack of 50
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17203754 50 Pack of 50
17233754 500 Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17203754 Supplier Neogen Petrifilm™ Supplier No. 7100039311
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
Designed to provide fast, accurate results in detecting coliform contamination.
  • Utilizes the convenience and efficiency of the Neogen Petrifilm™ Plate platform.
  • Proven cost effective tool for the detection of coliform.
  • Results in 4-24 hours.
  • This plate has been awarded Official Methods of Analysis (OMA) #2000.15 for Food and NF VALIDATION certificates: Neogen-01/5-03/97 A for all human food products , Neogen-01/5-03/97 B for All human food products and Neogen-01/5-03/97 C for All human food products (except process pork products) by AFNOR Certification.
  • Food processors and contract labs worldwide trust Neogen Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Product Type Count Plate
Test Type Environmental Test
Quantity 50