missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Rappaport-Vassiliadis (RV) Enrichment Broth (Bottled)

Product Code. 12796919
This item is not returnable. View return policy
Product Code. 12796919 Supplier Thermo Scientific™ Supplier No. BO0203E
  • ONLINE EXCLUSIVE PRICE
    66.75 €
    65.11 € / Pack of 24
    Save 1.64 €
    2% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively enrich Salmonella spp. from food and environmental samples using these ready prepared bottles.

Selectively enrich Salmonella spp. from food and environmental samples with Thermo Scientific™ Oxoid™ Rappaport-Vassiliadis (RV) Enrichment Broth ready prepared bottled medium. Rappaport-Vassiliadis (RV) Enrichment Broth selects for Salmonella species by exploiting their characteristics when compared with other Enterobacteriaceae, such as the ability to survive at relatively high osmotic pressure, multiply at relatively low pH values, be relatively more resistant to malachite green, and have relatively less demanding nutritional requirements1.

Remove the need for autoclaving and with convenient, pre-filled and ready prepared bottles of Rappaport-Vassiliadis Enrichment Broth.

  • Selective: malachite green, low pH and high magnesium chloride concentration inhibit many Gram-positive bacteria, while selectively enriching for salmonellae.
  • Superior performance: superior performance to Selenite Enrichment Broth and Tetrathionate Broth for enrichment of salmonellae with the exception of Salmonella typhi1.
  • Convenient: the pre-filled bottles are ready for immediate use.
  • Simple: easy-to-use with minimum training.
  • Long shelf life: up to 12 months.

It is important that the inoculum size used for enrichment culture in Rappaport- Vassiliadis Enrichment Broth is sufficiently small as not to interfere with its selectivity. Inoculum/broth ratios 1:100 to 1:2000 have been suggested2

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Blue, clear
Content And Storage 2 to 25°C
Description Rappaport- Vassiliadis (RV) Enrichment Broth (Bottled)
Format Universal Bottles
Manufacturing Origin United Kingdom
Product Type Microbiology Media
Fill Weight 10.0g + 0.3g
Packaging Type Twenty four universals with screw cap closures in a box
Quantity 24/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.