missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Rappaport-Vassiliadis Soya (RVS) Peptone Broth

Product Code. 11934042
Change view
Click to view available options
Fill Weight:
10 ± 0.5mL
Format:
Tube
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Format Unit Size
11934042 10 ± 0.5mL Tube Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11934042 Supplier Thermo Scientific™ Supplier No. TV5036E
  • ONLINE EXCLUSIVE PRICE
    67.70 €
    66.05 € / Pack of 50
    Save 1.65 €
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively enrich for the isolation of Salmonella species with Thermo Scientific™ Rappaport-Vassiliadis Soya Peptone Broth prepared media tubes.

Selectively enrich for the isolation of salmonellae from food and environmental specimens with Thermo Scientific™ Rappaport-Vassiliadis Soya Peptone Broth prepared media tubes. RVS Peptone Broth exploits characteristics of Salmonella species when compared with other Enterobacteriaceae, such as the ability to survive at relatively high osmotic pressure, multiply at relatively low pH values, be relatively more resistant to malachite green, and have relatively less demanding nutritional requirements.

RVS Peptone Broth is a modification of the Rappaport-Vassiliadis (RV) Enrichment Broth described earlier by van Schothorst and Renauld2. The modifications to their earlier formula are:

1. The addition of dipotassium hydrogen phosphate to buffer the medium so that the pH is maintained

during storage of the prepared broth.

2. Clarifying the optimum concentration of magnesium chloride 6H2O.

The two modifications are said to enhance the reliability of the enrichment broth1. Peterz et al.3 have also

highlighted the importance of the concentration of magnesium chloride in the final medium.

RVS Peptone Broth is based on the revised formulation and is recommended as the selective enrichment medium for the isolation of salmonellae from food and environmental specimens.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

 

TRUSTED_SUSTAINABILITY

Specifications

Color Pearl gentian blue, transparent
Content And Storage 2 to 25°C
Description Rappaport-Vassiliadis Soya(RVS) Peptone Broth
Format Tube
Manufacturing Origin Germany
Product Type Microbiology Media
Fill Volume 10 mL
Fill Weight 10 ± 0.5mL
Packaging Type 50 tubes in a carton box
Quantity 50/Pk.

For Laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.