missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAR alpha/NR1B1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
251.00 € - 549.00 €
Specifications
| Antigen | RAR alpha/NR1B1 |
|---|---|
| Dilution | Western Blot 1:750, ELISA 1:50, Chromatin Immunoprecipitation (ChIP) 4ug/ChIP, Chromatin Immunoprecipitation Sequencing 4ug/ChIP |
| Applications | Western Blot, ELISA, ChIP Assay, ChIP sequencing (ChIP-seq) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633278
|
Novus Biologicals
NBP3-11869-100ul |
100 μg |
549.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18691297
|
Novus Biologicals
NBP3-11869-25ul |
25 μg |
251.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAR alpha/NR1B1 Polyclonal antibody specifically detects RAR alpha/NR1B1 in Human samples. It is validated for Western Blot, ELISA, ChIP assay (ChIP), ChIP assay SequencingSpecifications
| RAR alpha/NR1B1 | |
| Western Blot, ELISA, ChIP Assay, ChIP sequencing (ChIP-seq) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Vision | |
| Whole antiserum | |
| 5914 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:750, ELISA 1:50, Chromatin Immunoprecipitation (ChIP) 4ug/ChIP, Chromatin Immunoprecipitation Sequencing 4ug/ChIP | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| alpha polypeptide, RAR-alpha, retinoic acid receptor, alpha | |
| Polyclonal antibody raised in rabbit against human RAR alpha/NR1B1 using two KLH-conjugated synthetic peptides containing sequences from the C-terminal region of the protein (Uniprot#: P10276). | |
| Whole Antiserum |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title