Learn More
Invitrogen™ PKC zeta Recombinant Rabbit Monoclonal Antibody (24GB4195)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556938
 View more versions of this product
Product Code. 30276029
  • 500.00 € / 100µL
Description
PKC zeta Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The PKC family of serine/threonine kinases, including PRKCZ (PKC zeta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. PKC zeta plays a key regulatory role in a variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PKC zeta
Recombinant Monoclonal
Unconjugated
PRKCZ
14 - 3 - 3 - zeta isoform; 14-3-3-zetaisoform; AI098070; aPKCzeta; atypical protein kinase C; C80388; kinase nPKC-zeta; KPCZ; nPKC-zeta; nPKC-zeta PRKCZ; PKC2; Pkcz; PKC-ZETA; Prkcz; protein kinase C zeta; protein kinase C zeta subspecies; protein kinase C zeta type; protein kinase C, zeta; r14-3-3; R74924; zetaPKC
Rabbit
Affinity chromatography
RUO
18762, 25522, 5590
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4195
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P09217, Q02956, Q05513
PRKCZ
A synthesized peptide derived from human PKC zeta (aa 350-592).
100 μL
Primary
Human, Mouse, Rat