missing translation for 'onlineSavingsMsg'
Learn More

Rare Earth and 'Geo' Elements Standard: Ba, Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Rb, Sc, Sm, Sr, Tb, Th, Tm, U, Y, Yb @ 100 μg/mL in 5% HNO3, VHG™

Product Code. 17927739
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17927739 100 mL 100mL
17958959 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17927739 Supplier VHG Labs Supplier No. VHGSM60A100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Rare Earth and 'Geo' Elements Standard: Ba, Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Rb, Sc, Sm, Sr, Tb, Th, Tm, U, Y, Yb @ 100 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lutetium/Barium/Cerium/Praseodymium/Terbium/Dysprosium/Europium/Gadolinium/Holmium/Neodymium/Rubidium/Samarium/Scandium/Thorium/Thulium/Uranium/Yttrium/Erbium/Lanthanum/Strontium/Ytterbium
CAS 7439-94-3, 7440-39-3, 7440-45-1, 7440-10-0, 7440-27-9, 7429-91-6
Methods ICP/ICP-MS
Analyte or Component Names Lutetium/Barium/Cerium/Praseodymium/Terbium/Dysprosium/Europium/Gadolinium/Holmium/Neodymium/Rubidium/Samarium/Scandium/Thorium/Thulium/Uranium/Yttrium/Erbium/Lanthanum/Strontium/Ytterbium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Lutetium 99.92 μg/mL/Barium 99.95 μg/mL/Cerium 99.97 μg/mL/Praseodymium 99.97 μg/mL/Terbium 99.97 μg/mL/Dysprosium 100 μg/mL/Europium 100 μg/mL/Gadolinium 100 μg/mL/Holmium 100 μg/mL/Neodymium 100 μg/mL/Rubidium 100 μg/mL/Samarium 100 μg/mL/Scandium 100 μg/mL/Thorium 100 μg/mL/Thulium 100 μg/mL/Uranium 100 μg/mL/Yttrium 100 μg/mL/Erbium 100.1 μg/mL/Lanthanum 100.1 μg/mL/Strontium 100.1 μg/mL/Ytterbium 100.1 μg/mL
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.