missing translation for 'onlineSavingsMsg'
Learn More

RASAL1 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

188.00 € - 470.00 €

Specifications

Antigen RASAL1
Dilution Western Blot 1:1000 - 1:5000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18607091
View Documents
Novus Biologicals
NBP2-94543-0.02ml
0.02 mL
188.00 €
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18646180
View Documents
Novus Biologicals
NBP2-94543-0.1ml
0.1 mL
470.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RASAL1 Polyclonal antibody specifically detects RASAL1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

RASAL1
Western Blot
Unconjugated
Rabbit
Signal Transduction
PBS (pH 7.3), 50% glycerol
8437
IgG
Affinity purified
Western Blot 1:1000 - 1:5000
Polyclonal
Purified
RUO
Human, Mouse, Rat
RAS protein activator like 1 (GAP1 like), RASALGAP1 like protein, rasGAP-activating-like protein 1
Recombinant fusion protein containing a sequence corresponding to amino acids 120-340 of human RASAL1 (NP_004649.2). ICLSVQMLEDGQGRCLRCHVLQARDLAPRDISGTSDPFARVFWGSQSLETSTIKKTRFPHWDEVLELREMPGAPSPLRVELWDWDMVGKNDFLGMVEFSPKTLQQKPPKGWFRLLPFPRAEEDSGGNLGALRVKVRLIEDRVLPSQCYQPLMELLMESVQGPAEEDTASPLALLEELTLGDCRQDLATKLVKLFLGRGLAGRFLDYLTRREVARTMDPNTL
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.