missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RASEF Recombinant Rabbit Monoclonal Antibody (K01_1X43)

Product Code. p-200114746
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826951 100 μL
30830432 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826951 Supplier Invitrogen™ Supplier No. MA563566

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: PETHDSEEDE GDED.

Binds predominantly GDP, and also GTP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASEF
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1X43
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene RASEF
Gene Accession No. Q5RI75, Q8IZ41
Gene Alias BC030882; RAB45; RAS and EF hand domain containing; RAS and EF-hand domain containing; ras and EF-hand domain-containing protein; Ras and EF-hand domain-containing protein homolog; RASEF; ras-related protein Rab-45
Gene Symbols RASEF
Host Species Rabbit
Immunogen A synthetic peptide of human RAB45 (aa 97-110).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158158, 242505
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.