missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl2l11, Rat, Clone: 14A8, Abnova™
Rat monoclonal antibody raised against recombinant Bcl2l11.
Brand: Abnova MAB3704.100ug
This item is not returnable.
View return policy
Specifications
| Bcl2l11 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Bcl2l11 | |
| Bcl2l11 | |
| Recombinant protein corresponding to mouse Bcl2l11. | |
| RUO | |
| 12125 | |
| Human, Monkey, Mouse | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 14A8 | |
| Rat monoclonal antibody raised against recombinant Bcl2l11. | |
| In PBS (0.02% sodium azide) | |
| 1500006F24Rik/Bim/Bod | |
| Rat | |
| 100 μg | |
| Primary | |
| Recognizes human, mouse and monkey BimL, BimEL and BimS. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction