missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL24/Eotaxin-2/MPIF-2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
214.00 € - 2055.00 €
Specifications
| Antigen | CCL24/Eotaxin-2/MPIF-2 |
|---|---|
| Clone | 106521 |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.06-0.3 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18770048
![]() |
R&D Systems
MAB528-SP |
25 μg |
214.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18777408
![]() |
R&D Systems
MAB528-100 |
100 μg |
517.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18784037
![]() |
R&D Systems
MAB528-500 |
500 μg |
2055.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL24/Eotaxin-2/MPIF-2 Monoclonal specifically detects CCL24/Eotaxin-2/MPIF-2 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| CCL24/Eotaxin-2/MPIF-2 | |
| Western Blot 1 ug/mL, Neutralization 0.06-0.3 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Q9JKC0 | |
| 6369 | |
| E. coli-derived recombinant mouse CCL24/Eotaxin-2/MPIF-2 Ile24-Val119 Accession # Q9JKC0 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CCL24/Eotaxin-2/MPIF-2 in Western blots. In Western blots, no cross-reactivity with recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 28, recombinant mouse CCL1, 2, 3, 4, 6, 7, 9/10/MIP-1 gamma , 11, 12, 19, 20, 21, 22, 25, 28, or recombinant rat CCL20 is observed. |
| 106521 | |
| Western Blot, Neutralization, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, member 24, MPIF2, MPIF-2, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2Small-inducible cytokine A24, SCYA24CK-beta-6 | |
| CCL24 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title