missing translation for 'onlineSavingsMsg'
Learn More

Rat B7-2/CD86 Antibody, R&D Systems™

Product Code. 18058921
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18058921 100 μg 100µg
18716138 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18058921 Supplier R&D Systems Supplier No. AF1340

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

B7-2/CD86 Polyclonal specifically detects B7-2/CD86 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen B7-2/CD86
Applications Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Gene Accession No. O35531
Gene Alias Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86
Gene Symbols CD86
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat B7-2/CD86 Val29-Ile250 Accession # O35531
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 942
Test Specificity Detects rat B7-2/CD86 in ELISAs and Western blots. In sandwich immunoassays, less than 20% cross-reactivity with recombinant mouse (rm) B7-2 is observed and less than 0.2% cross-reactivity with recombinant rat B7-1, rmB7-1, and recombinant human B7-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.