missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat CCR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
222.00 € - 500.00 €
Specifications
| Antigen | CCR2 |
|---|---|
| Clone | 890231R |
| Dilution | Flow Cytometry 0.25 μg/106 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18724087
![]() |
R&D Systems
MAB8368R-SP |
25 μg |
222.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18724257
![]() |
R&D Systems
MAB8368R-100 |
100 μg |
500.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR2 Monoclonal specifically detects CCR2 in Rat samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| CCR2 | |
| Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Rat | |
| O55193 | |
| 729230.0 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 890231R | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
| NS0 mouse myeloma cell line transfected with rat CCR2, Accession No. O55193 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects rat CCR2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title