missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat CCR6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8320R-100
This item is not returnable.
View return policy
Description
CCR6 Monoclonal specifically detects CCR6 in Rat samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CCR6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKRL3, CKR-L3, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPRCY4, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22 | |
| NS0 mouse myeloma cell line transfected with rat CCR6, Met1-Met366, Accession No. Q5BK58 | |
| 100 μg | |
| 1235.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 876515R | |
| Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| Q5BK58 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects rat CCR6 in direct ELISAs. | |
| Rat | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction